Product Details

SNP ID
rs9324066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103928513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGACGCCTGGGCCTTGAGGATG[C/T]TGCGGAAGCTCACCATCGAGCAGAT
Phenotype
MIM: 604573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf2 PubMed Links

Gene Details

Gene
C14orf2
Gene Name
chromosome 14 open reading frame 2
There are no transcripts associated with this gene.

Gene
TDRD9
Gene Name
tudor domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153046.2 171 Silent Mutation CTG,TTG L2L NP_694591.2
XM_005267309.3 171 Missense Mutation CTG,TTG L2L XP_005267366.1
XM_006720019.3 171 Missense Mutation CTG,TTG L2L XP_006720082.1
XM_006720020.3 171 Missense Mutation CTG,TTG L2L XP_006720083.1
XM_011536397.2 171 Missense Mutation CTG,TTG L2L XP_011534699.1
XM_011536398.2 171 Intron XP_011534700.1
XM_011536400.2 171 Silent Mutation CTG,TTG L2L XP_011534702.1
XM_011536401.2 171 Intron XP_011534703.1
XM_011536402.2 171 Intron XP_011534704.1
XM_017020955.1 171 UTR 5 XP_016876444.1

View Full Product Details