Product Details

SNP ID
rs9940751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58402032 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAATTACATGCATGTTGAACTTT[G/T]TCTTTCTTTTACACCTTATTTCATA
Phenotype
MIM: 610610
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GINS3 PubMed Links

Gene Details

Gene
GINS3
Gene Name
GINS complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126129.1 Intron NP_001119601.1
NM_001126130.1 Intron NP_001119602.1
NM_022770.3 Intron NP_073607.2

View Full Product Details