Product Details
- SNP ID
-
rs6136921
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:20035010 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCCAGGTTAAAGGAAAACTCTCA[G/T]ATCGGTTAATGCAGTTGAATTTTTG
- Phenotype
-
MIM: 610952
MIM: 610833
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CRNKL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11467884] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRNKL1
- Gene Name
- crooked neck pre-mRNA splicing factor 1
- Gene
- NAA20
- Gene Name
- N(alpha)-acetyltransferase 20, NatB catalytic subunit
There are no transcripts associated with this gene.
View Full Product Details