Product Details

SNP ID
rs8048728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89211456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCCCCTGCTGCCCCAGGTCTCC[A/G]GGAAAACGCAGACTGGGCACCCCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC22A31 PubMed Links

Gene Details

Gene
SLC22A31
Gene Name
solute carrier family 22 member 31
There are no transcripts associated with this gene.

Gene
ZNF778
Gene Name
zinc finger protein 778
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201407.1 Intron NP_001188336.1
NM_182531.3 Intron NP_872337.2
XM_005256288.2 Intron XP_005256345.1
XM_011522940.2 Intron XP_011521242.1
XM_017023015.1 Intron XP_016878504.1
XM_017023016.1 Intron XP_016878505.1
XM_017023017.1 Intron XP_016878506.1

View Full Product Details