Product Details

SNP ID
hCV30433848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36256194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGATATGTCAATAGATGTTACTT[A/T]AAAAAGATGTGGTCAGTCAGGCATG
Phenotype
MIM: 612121
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PNPLA1 PubMed Links

Gene Details

Gene
PNPLA1
Gene Name
patatin like phospholipase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details