Product Details

SNP ID
rs10052079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115829683 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGACTTGAGAGTTTAAATTAAAA[A/G]TAAATTGTGTTGTAGGTTCTAGTCA
Phenotype
MIM: 609608
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG12 PubMed Links

Gene Details

Gene
ATG12
Gene Name
autophagy related 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277783.1 2706 UTR 3 NP_001264712.1
NM_004707.3 2706 UTR 3 NP_004698.3

View Full Product Details