Product Details

SNP ID
rs5937763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:74084101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATAACACAGGGAAAAGTTAAAT[C/G]CTAAGCCAAAATATGCTCCAACATA
Phenotype
MIM: 300936
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FTX PubMed Links

Gene Details

Gene
FTX
Gene Name
FTX transcript, XIST regulator (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details