Product Details

SNP ID
rs6576088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105412860 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGCTGGACCTGTGCGGTGGCCTG[C/G]ACCACCAGATACACTACATGCTGGC
Phenotype
MIM: 603526
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100507437 PubMed Links

Gene Details

Gene
LOC100507437
Gene Name
uncharacterized LOC100507437
There are no transcripts associated with this gene.

Gene
MTA1
Gene Name
metastasis associated 1
There are no transcripts associated with this gene.

Gene
TEX22
Gene Name
testis expressed 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195082.1 1846 UTR 3 NP_001182011.1
XM_006720234.3 1846 UTR 3 XP_006720297.1

View Full Product Details