Product Details

SNP ID
rs13263962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:120418359 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCATGTGCATGTGTATCTTGAAG[A/G]TGTATTTTTATAAATGAGATTGCTG
Phenotype
MIM: 610200
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL13 PubMed Links

Gene Details

Gene
MRPL13
Gene Name
mitochondrial ribosomal protein L13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014078.5 Intron NP_054797.2

View Full Product Details