Product Details

SNP ID
rs10915949
Assay Type
Functionally tested
NCBI dbSNP Submissions
57
Location
Chr.1:226152063 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCATATTTGGCTTTTCAACACACA[A/C]GGTTAGAAGTAAAATAGTATTCAGA
Phenotype
MIM: 606809
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACBD3 PubMed Links

Gene Details

Gene
ACBD3
Gene Name
acyl-CoA binding domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022735.3 Intron NP_073572.2

View Full Product Details