Product Details

SNP ID
rs12841526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:45158170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGGGGTGGGAGGCCCCCTCCCC[C/G]ACCACAGGCTACACTCGGGGCGGGG
Phenotype
MIM: 300959
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CXorf36 PubMed Links

Gene Details

Gene
CXorf36
Gene Name
chromosome X open reading frame 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024689.2 Intron NP_078965.2
NM_176819.3 Intron NP_789789.2
XM_005272670.1 Intron XP_005272727.1
XM_006724559.1 Intron XP_006724622.1

View Full Product Details