Product Details

SNP ID
rs13170723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73552605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATTAATTTATAATTTTAAATAT[A/C]CAGTAAAACATAGTTATAAAAAGAG
Phenotype
MIM: 605787
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRA2 PubMed Links

Gene Details

Gene
ANKRA2
Gene Name
ankyrin repeat family A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023039.4 1651 UTR 3 NP_075526.1
XM_005248560.2 1651 UTR 3 XP_005248617.1
XM_017009678.1 1651 UTR 3 XP_016865167.1

View Full Product Details