Product Details

SNP ID
rs12757356
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:151180359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTCTGGAAACCCTTCCAATGAT[G/T]GAACCTCTCTTGCACTTGAACTGTT
Phenotype
MIM: 605834 MIM: 600607
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMOD4 PubMed Links

Gene Details

Gene
TMOD4
Gene Name
tropomodulin 4
There are no transcripts associated with this gene.

Gene
VPS72
Gene Name
vacuolar protein sorting 72 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271087.1 Intron NP_001258016.1
NM_001271088.1 Intron NP_001258017.1
NM_005997.2 Intron NP_005988.1
XM_017002205.1 Intron XP_016857694.1

View Full Product Details