Product Details

SNP ID
rs11128020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:88053143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTACACCTCCAAAGCAGCATATG[A/C]CTCTTAGACATCCTCATTTGTTATC
Phenotype
MIM: 603363
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CGGBP1 PubMed Links

Gene Details

Gene
CGGBP1
Gene Name
CGG triplet repeat binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008390.1 3327 UTR 3 NP_001008391.1
NM_001195308.1 3327 UTR 3 NP_001182237.1
NM_003663.3 3327 UTR 3 NP_003654.3
XM_017007355.1 3327 UTR 3 XP_016862844.1
Gene
ZNF654
Gene Name
zinc finger protein 654
There are no transcripts associated with this gene.

View Full Product Details