Product Details

SNP ID
rs12839420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:7148031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACAAGACCGCCCTTACTGGAG[A/G]CGGCGGCTGCACACTACCCACCCAG
Phenotype
MIM: 306480 MIM: 300747
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4767 PubMed Links

Gene Details

Gene
MIR4767
Gene Name
microRNA 4767
There are no transcripts associated with this gene.

Gene
PUDP
Gene Name
pseudouridine 5'-phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135565.1 139 Intron NP_001129037.1
NM_001178135.1 139 Intron NP_001171606.1
NM_001178136.1 139 Intron NP_001171607.1
NM_012080.4 139 Intron NP_036212.3
Gene
STS
Gene Name
steroid sulfatase (microsomal), isozyme S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000351.5 139 Intron NP_000342.2
NM_001320750.1 139 Intron NP_001307679.1
NM_001320751.1 139 UTR 5 NP_001307680.1
NM_001320752.1 139 UTR 5 NP_001307681.1
NM_001320753.1 139 Intron NP_001307682.1
NM_001320754.1 139 Intron NP_001307683.1

View Full Product Details