Product Details
- SNP ID
-
rs10794665
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
64
- Location
-
Chr.1:24119638 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTCTTTCCCACCCCAGACCCCTCT[A/G]GCTTTGCCTGTCCTGAGGAAATCCA
- Phenotype
-
MIM: 605457
MIM: 616832
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IL22RA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77639105] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL22RA1
- Gene Name
- interleukin 22 receptor subunit alpha 1
- Gene
- MYOM3
- Gene Name
- myomesin 3
There are no transcripts associated with this gene.
View Full Product Details