Product Details

SNP ID
rs13402301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:234493473 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTACAGAAATCAACTGAGACTCT[C/T]AACTAGTAGTTGATACACCACAGGG
Phenotype
MIM: 604787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL4C PubMed Links

Gene Details

Gene
ARL4C
Gene Name
ADP ribosylation factor like GTPase 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282431.1 3406 UTR 3 NP_001269360.1
NM_005737.3 3406 UTR 3 NP_005728.2

View Full Product Details