Product Details

SNP ID
rs13256679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143428621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCCACCCGACTGAGCACAGGA[C/T]TCACCCCCCTCCCCCCACCCCCGGT
Phenotype
MIM: 610303
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAFA PubMed Links

Gene Details

Gene
MAFA
Gene Name
MAF bZIP transcription factor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201589.3 1812 UTR 3 NP_963883.2
Gene
MAFA-AS1
Gene Name
MAFA antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZC3H3
Gene Name
zinc finger CCCH-type containing 3
There are no transcripts associated with this gene.

View Full Product Details