Product Details

SNP ID
rs11102279
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:108713513 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTCCAAACTTTCCTTTTCAGGT[C/T]GCTTCAGCAAAATCAGGTACAATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FNDC7 PubMed Links

Gene Details

Gene
FNDC7
Gene Name
fibronectin type III domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144937.1 66 Silent Mutation GTC,GTT V22V NP_001138409.1

View Full Product Details