Product Details

SNP ID
rs12459058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46770881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTACATAACAGCCTACCACGGAT[C/T]ATGGCATAGTAGAAAGCATTTAAAA
Phenotype
MIM: 109190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC1A5 PubMed Links

Gene Details

Gene
SLC1A5
Gene Name
solute carrier family 1 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145144.1 Intron NP_001138616.1
NM_001145145.1 Intron NP_001138617.1
NM_005628.2 Intron NP_005619.1
XM_005259167.4 Intron XP_005259224.1

View Full Product Details