Product Details

SNP ID
rs12494412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:161222142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCACTATGGAGAACGCTTGAGGG[G/T]GGGTGGGAAAGAGCGTATATTGTCA
Phenotype
MIM: 611021
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NMD3 PubMed Links

Gene Details

Gene
NMD3
Gene Name
NMD3 ribosome export adaptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320227.1 Intron NP_001307156.1
NM_015938.4 Intron NP_057022.2
XM_005247511.1 Intron XP_005247568.1
XM_005247512.1 Intron XP_005247569.1

View Full Product Details