Product Details
- SNP ID
-
rs11023241
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:14507046 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGGTGAGACCAGCAAAGAACCATG[A/T]AGGTGAGCCCCAGTCAGCCGCAAAA
- Phenotype
-
MIM: 600959
MIM: 602854
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
COPB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112296502] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COPB1
- Gene Name
- coatomer protein complex subunit beta 1
There are no transcripts associated with this gene.
- Gene
- PSMA1
- Gene Name
- proteasome subunit alpha 1
View Full Product Details