Product Details

SNP ID
rs11018912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:90209477 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTCTATCATTCATTCTACACGCA[C/T]TGTTCTAAGGATTCAAGATACAACA
Phenotype
MIM: 604353
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHORDC1 PubMed Links

Gene Details

Gene
CHORDC1
Gene Name
cysteine and histidine rich domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144073.1 Intron NP_001137545.1
NM_012124.2 Intron NP_036256.2
XM_017017540.1 Intron XP_016873029.1
XM_017017541.1 Intron XP_016873030.1

View Full Product Details