Product Details

SNP ID
rs12778602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112427720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACCCTCTGTGCCTTCTTCTTTG[G/T]TTTGTGATAAACATAACTTGCCAAC
Phenotype
MIM: 605677
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACSL5 PubMed Links

Gene Details

Gene
ACSL5
Gene Name
acyl-CoA synthetase long-chain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016234.3 2555 UTR 3 NP_057318.2
NM_203379.1 2555 UTR 3 NP_976313.1
NM_203380.1 2555 UTR 3 NP_976314.1
XM_017016324.1 2555 UTR 3 XP_016871813.1
Gene
ZDHHC6
Gene Name
zinc finger DHHC-type containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303134.1 2555 Intron NP_001290063.1
NM_022494.2 2555 Intron NP_071939.1
XM_005270051.4 2555 Intron XP_005270108.1
XM_006717948.3 2555 Intron XP_006718011.1
XM_006717949.3 2555 Intron XP_006718012.1
XM_011540091.2 2555 Intron XP_011538393.1
XM_011540092.2 2555 Intron XP_011538394.1
XM_011540093.1 2555 Intron XP_011538395.1
XM_011540094.2 2555 Intron XP_011538396.1
XM_011540095.2 2555 Intron XP_011538397.1
XM_017016561.1 2555 Intron XP_016872050.1
XM_017016562.1 2555 Intron XP_016872051.1
XM_017016563.1 2555 Intron XP_016872052.1
XM_017016564.1 2555 Intron XP_016872053.1
XM_017016565.1 2555 Intron XP_016872054.1
XM_017016566.1 2555 Intron XP_016872055.1
XM_017016567.1 2555 Intron XP_016872056.1
XM_017016568.1 2555 Intron XP_016872057.1
XM_017016569.1 2555 Intron XP_016872058.1

View Full Product Details