Product Details

SNP ID
rs12259524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97575185 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCTGTAGGGTCTGAGGAAGATGT[G/T]GGTGAGCATTAGAGTCCCCAGTTGT
Phenotype
MIM: 610734 MIM: 613597 MIM: 616388
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD2 PubMed Links

Gene Details

Gene
ANKRD2
Gene Name
ankyrin repeat domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129981.2 Intron NP_001123453.1
NM_001291218.1 Intron NP_001278147.1
NM_001291219.1 Intron NP_001278148.1
NM_020349.3 Intron NP_065082.2
Gene
HOGA1
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
There are no transcripts associated with this gene.

Gene
UBTD1
Gene Name
ubiquitin domain containing 1
There are no transcripts associated with this gene.

View Full Product Details