Product Details

SNP ID
rs7150226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49788874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGCCTCTCTGTCTTAATTTAA[C/T]TCAGTTCCCATCAGAATCAAAGCCT
Phenotype
MIM: 611280 MIM: 608378
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHDC2 PubMed Links

Gene Details

Gene
KLHDC2
Gene Name
kelch domain containing 2
There are no transcripts associated with this gene.

Gene
NEMF
Gene Name
nuclear export mediator factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011537317.2 Intron XP_011535619.1
XM_011537318.2 Intron XP_011535620.1
XM_017021760.1 Intron XP_016877249.1
XM_017021761.1 Intron XP_016877250.1

View Full Product Details