Product Details

SNP ID
rs12371092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51393245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCTGCTTCAGCCTCCACAGAT[A/G]CATGCCACCATGCCCAGCTAACTTT
Phenotype
MIM: 605148 MIM: 605024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALNT6 PubMed Links

Gene Details

Gene
GALNT6
Gene Name
polypeptide N-acetylgalactosaminyltransferase 6
There are no transcripts associated with this gene.

Gene
SLC4A8
Gene Name
solute carrier family 4 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039960.2 Intron NP_001035049.1
NM_001258401.2 Intron NP_001245330.1
NM_001258402.1 Intron NP_001245331.1
NM_001258403.1 Intron NP_001245332.1
NM_001267615.1 Intron NP_001254544.1
XM_006719700.1 Intron XP_006719763.1
XM_011539010.1 Intron XP_011537312.1
XM_011539011.1 Intron XP_011537313.1
XM_011539014.2 Intron XP_011537316.1
XM_017020240.1 Intron XP_016875729.1
XM_017020241.1 Intron XP_016875730.1
XM_017020242.1 Intron XP_016875731.1

View Full Product Details