Product Details
- SNP ID
-
rs11084060
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51212754 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACAATGCACAGCGGAGTCTGTCCT[C/T]AGATGCCCGTCCTCCTCCCCAGAAT
- Phenotype
-
MIM: 159590
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD33
PubMed Links
Gene Details
- Gene
- CD33
- Gene Name
- CD33 molecule
- Gene
- MIR8074
- Gene Name
- microRNA 8074
There are no transcripts associated with this gene.
View Full Product Details