Product Details

SNP ID
rs12042224
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:244414450 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTTTAAAAGCATATTGGAGATG[C/T]TGATCAGAATCATTTACCTCTTCCA
Phenotype
MIM: 103060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADSS PubMed Links

Gene Details

Gene
ADSS
Gene Name
adenylosuccinate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126.3 Intron NP_001117.2
XM_011544108.2 Intron XP_011542410.1

View Full Product Details