Product Details

SNP ID
rs11263687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:22788625 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACACCAGATGTAAGACATTAGGGA[C/A]AAACGAAGGAAAGCTGAATACCAGC
Phenotype
MIM: 608145
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
LOC283683 PubMed Links

Gene Details

Gene
LOC283683
Gene Name
uncharacterized LOC283683
There are no transcripts associated with this gene.

Gene
NIPA1
Gene Name
non imprinted in Prader-Willi/Angelman syndrome 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142275.1 Intron NP_001135747.1
NM_144599.4 Intron NP_653200.2

View Full Product Details