Product Details

SNP ID
rs12406788
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:204039426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACACAGTGGGCACCACTTGCCG[C/T]TGGGTAGAGGCAAGAGGCATTGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00303 PubMed Links

Gene Details

Gene
LINC00303
Gene Name
long intergenic non-protein coding RNA 303
There are no transcripts associated with this gene.

View Full Product Details