Product Details

SNP ID
rs11770506
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:55022686 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGAAAGGACACCAGTGGTTTGTG[C/T]GTTAGACCCACACTGCCGTAGCACA
Phenotype
MIM: 131550
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
EGFR PubMed Links
Additional Information
For this assay, SNP(s) [rs116855255] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
EGFR
Gene Name
epidermal growth factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005228.3 Intron NP_005219.2
NM_201282.1 Intron NP_958439.1
NM_201283.1 Intron NP_958440.1
NM_201284.1 Intron NP_958441.1

View Full Product Details