Product Details

SNP ID
rs12408947
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:234630684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGTGAGGCAAAGATGGTCTCCA[G/T]TACAACGCAGTGCCTGCTGCATAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00184 PubMed Links

Gene Details

Gene
LINC00184
Gene Name
long intergenic non-protein coding RNA 184
There are no transcripts associated with this gene.

View Full Product Details