Product Details

SNP ID
rs12086354
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:92301801 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATGCACTAATGAAAAATAAGTA[C/T]GTAACAAGGTGATGACACGTAATGT
Phenotype
MIM: 601749 MIM: 611476
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLMN PubMed Links

Gene Details

Gene
GLMN
Gene Name
glomulin, FKBP associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319683.1 Intron NP_001306612.1
NM_053274.2 Intron NP_444504.1
XM_005270401.3 Intron XP_005270458.1
XM_006710309.2 Intron XP_006710372.1
XM_011540546.2 Intron XP_011538848.1
XM_017000137.1 Intron XP_016855626.1
XM_017000138.1 Intron XP_016855627.1
XM_017000139.1 Intron XP_016855628.1
XM_017000140.1 Intron XP_016855629.1
XM_017000141.1 Intron XP_016855630.1
XM_017000142.1 Intron XP_016855631.1
XM_017000143.1 Intron XP_016855632.1
XM_017000144.1 Intron XP_016855633.1
Gene
RPAP2
Gene Name
RNA polymerase II associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024813.2 Intron NP_079089.2
XM_005271223.2 Intron XP_005271280.1
XM_011542167.2 Intron XP_011540469.1
XM_017002362.1 Intron XP_016857851.1
XM_017002363.1 Intron XP_016857852.1
XM_017002364.1 Intron XP_016857853.1

View Full Product Details