Product Details

SNP ID
rs10924874
Assay Type
Functionally tested
NCBI dbSNP Submissions
54
Location
Chr.1:246953000 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCTAGGAAAGGATGTGACCCTAC[C/T]TGAGCCACAATTTAGAAGGGCATGT
Phenotype
MIM: 616348
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF670-ZNF695 PubMed Links

Gene Details

Gene
ZNF670-ZNF695
Gene Name
ZNF670-ZNF695 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
ZNF695
Gene Name
zinc finger protein 695
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204221.1 Intron NP_001191150.1
NM_020394.4 Intron NP_065127.4

View Full Product Details