Product Details
- SNP ID
-
rs11878479
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:23096981 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGATATGACCCTTTTATTTTCCCG[G/T]TTCTGGCATATTCTGGGTATAGTAA
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF730
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78509685] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF730
- Gene Name
- zinc finger protein 730
View Full Product Details