Product Details

SNP ID
rs12595504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34345997 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCTGGAGGCTTCCAGACAGCCAC[A/G]GTTAGTGCCCAAACCTGAGAGGATG
Phenotype
MIM: 606471 MIM: 608963 MIM: 604878
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOP10 PubMed Links

Gene Details

Gene
NOP10
Gene Name
NOP10 ribonucleoprotein
There are no transcripts associated with this gene.

Gene
NUTM1
Gene Name
NUT midline carcinoma family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284292.1 444 Missense Mutation CAG,CGG Q21R NP_001271221.1
NM_001284293.1 444 Intron NP_001271222.1
NM_175741.2 444 UTR 5 NP_786883.1
Gene
SLC12A6
Gene Name
solute carrier family 12 member 6
There are no transcripts associated with this gene.

View Full Product Details