Product Details
- SNP ID
-
rs12900317
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:90240924 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTCAGCTCTATGGCTCTTCCACAT[C/G]ATTCAAACGAAACTTCCTATTTGCT
- Phenotype
-
MIM: 602293
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CIB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78054010] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CIB1
- Gene Name
- calcium and integrin binding 1
- Gene
- GDPGP1
- Gene Name
- GDP-D-glucose phosphorylase 1
- Gene
- TTLL13P
- Gene Name
- tubulin tyrosine ligase like 13, pseudogene
There are no transcripts associated with this gene.
View Full Product Details