Product Details

SNP ID
rs11755921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31903977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACCTAGGCTTAAAGCCACATTC[A/C]TAGGAGTTTCTGAGCCTACTCTGGC
Phenotype
MIM: 613927 MIM: 604599
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C2 PubMed Links

Gene Details

Gene
C2
Gene Name
complement component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000063.5 Intron NP_000054.2
NM_001145903.2 Intron NP_001139375.1
NM_001178063.2 Intron NP_001171534.1
NM_001282457.1 Intron NP_001269386.1
NM_001282458.1 Intron NP_001269387.1
NM_001282459.1 Intron NP_001269388.1
Gene
EHMT2
Gene Name
euchromatic histone lysine methyltransferase 2
There are no transcripts associated with this gene.

Gene
ZBTB12
Gene Name
zinc finger and BTB domain containing 12
There are no transcripts associated with this gene.

View Full Product Details