Product Details

SNP ID
rs13418607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:192759642 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATAGTATTATTCATTTAGAGGCT[C/T]GATATTATTATTATAAAATGCATGG
Phenotype
MIM: 605443
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCGEM1 PubMed Links

Gene Details

Gene
PCGEM1
Gene Name
PCGEM1, prostate-specific transcript (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details