Product Details

SNP ID
rs11666169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40031740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAAGTTGAATTATGTTGACGCT[A/T]TAATGGCATTCATTTCTCAATAGTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF780B PubMed Links

Gene Details

Gene
ZNF780B
Gene Name
zinc finger protein 780B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005851.2 5416 UTR 3 NP_001005851.1
XM_005258590.4 5416 UTR 3 XP_005258647.1
XM_005258591.4 5416 UTR 3 XP_005258648.1
XM_005258592.3 5416 UTR 3 XP_005258649.1
XM_005258593.2 5416 UTR 3 XP_005258650.1
XM_005258595.3 5416 UTR 3 XP_005258652.1
XM_006723072.3 5416 UTR 3 XP_006723135.1
XM_011526593.2 5416 UTR 3 XP_011524895.1
XM_017026425.1 5416 UTR 3 XP_016881914.1
XM_017026426.1 5416 UTR 3 XP_016881915.1
XM_017026427.1 5416 UTR 3 XP_016881916.1

View Full Product Details