Product Details

SNP ID
rs12347519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101421098 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAATACCCTCAAACTTGAAATTTA[G/T]ATTGGTAGTTCTCAAACTTGACTGT
Phenotype
MIM: 612724
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDOB PubMed Links

Gene Details

Gene
ALDOB
Gene Name
aldolase, fructose-bisphosphate B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000035.3 1888 UTR 3 NP_000026.2

View Full Product Details