Product Details

SNP ID
rs11190499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100379325 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATAGGAATCACGGTAGTAGCACCA[A/C]ACTGTACCAGTGGGCATTACATTCC
Phenotype
MIM: 616240
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OLMALINC PubMed Links

Gene Details

Gene
OLMALINC
Gene Name
oligodendrocyte maturation-associated long intergenic non-coding RNA
There are no transcripts associated with this gene.

View Full Product Details