Product Details

SNP ID
rs12250159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93761093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAATAAACTCATGTGCGGTGTGTA[A/G]GAAATATATGAGAAGGCTCTGAATA
Phenotype
MIM: 604619
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LGI1 PubMed Links

Gene Details

Gene
LGI1
Gene Name
leucine rich glioma inactivated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308275.1 Intron NP_001295204.1
NM_001308276.1 Intron NP_001295205.1
NM_005097.3 Intron NP_005088.1
XM_017016911.1 Intron XP_016872400.1
XM_017016912.1 Intron XP_016872401.1
Gene
LOC101927032
Gene Name
uncharacterized LOC101927032
There are no transcripts associated with this gene.

View Full Product Details