Product Details

SNP ID
rs13412209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:236538241 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAATCCCTGACTTGATTAACTGT[G/T]ATTGAGTGTGTTCTGCAAAGTAACA
Phenotype
MIM: 610376
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACKR3 PubMed Links

Gene Details

Gene
ACKR3
Gene Name
atypical chemokine receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020311.2 Intron NP_064707.1
XM_005246097.2 Intron XP_005246154.1
XM_005246098.3 Intron XP_005246155.1
XM_017004516.1 Intron XP_016860005.1

View Full Product Details