Product Details
- SNP ID
-
rs9333326
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:43157519 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAGTGGCAGCACCGGAATCTGCTG[A/G]ATGCCTGACACTCTGTTCCTTTAAA
- Phenotype
-
MIM: 109610
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TSPO
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138912] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TSPO
- Gene Name
- translocator protein
- Gene
- TTLL12
- Gene Name
- tubulin tyrosine ligase like 12
There are no transcripts associated with this gene.
View Full Product Details