Product Details

SNP ID
rs11906023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62388964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACATAACTGAAGGGAAGGCTTC[A/G]CACATCACAAAGACCGGGTTGGTTA
Phenotype
MIM: 180477
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CABLES2 PubMed Links

Gene Details

Gene
CABLES2
Gene Name
Cdk5 and Abl enzyme substrate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031215.2 3451 UTR 3 NP_112492.2
Gene
RPS21
Gene Name
ribosomal protein S21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024.3 3451 Intron NP_001015.1

View Full Product Details