Product Details

SNP ID
rs11159269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77509982 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTAGCAGGTAAGTTCATTGCTTA[C/T]AAGTTTGTGACTTTTACAAACCCTA
Phenotype
MIM: 605713
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPTLC2 PubMed Links

Gene Details

Gene
SPTLC2
Gene Name
serine palmitoyltransferase long chain base subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004863.3 4179 UTR 3 NP_004854.1
XM_011537384.2 4179 Intron XP_011535686.1

View Full Product Details