Product Details

SNP ID
rs12886690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:90065890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGCTTCAGATTTGGTTTCAGGC[C/T]TCATGGCAGACAGAGTCATGACCCA
Phenotype
MIM: 607367
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNK13 PubMed Links

Gene Details

Gene
KCNK13
Gene Name
potassium two pore domain channel subfamily K member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022054.3 Intron NP_071337.2

View Full Product Details