Product Details

SNP ID
rs12923869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3443730 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTGAGCTCCGGGCCTGTTTGCCTG[C/T]TGAAGTAGAGTCTTAGGGTGACCCC
Phenotype
MIM: 614246 MIM: 614685
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAA60 PubMed Links

Gene Details

Gene
NAA60
Gene Name
N(alpha)-acetyltransferase 60, NatF catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083600.2 120 Intron NP_001077069.1
NM_001083601.2 120 UTR 5 NP_001077070.1
NM_001317093.1 120 UTR 5 NP_001304022.1
NM_001317094.1 120 Intron NP_001304023.1
NM_001317095.1 120 Intron NP_001304024.1
NM_001317096.1 120 Intron NP_001304025.1
NM_001317097.1 120 Intron NP_001304026.1
NM_001317098.1 120 Intron NP_001304027.1
NM_024845.3 120 Intron NP_079121.1
Gene
ZNF597
Gene Name
zinc finger protein 597
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152457.2 120 Intron NP_689670.1

View Full Product Details